missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycophenolate mofetil, 98%, Thermo Scientific Chemicals
238.88 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 95.0°C to 96.0°C |
Ignition Residue | 0.1% max |
Loss on Drying | 0.5% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 433.5 |
Percent Purity | 98% |
Chemical Name or Material | Mycophenolate mofetil |
Safety and Handling
Product Identifier
- Mycophenolate mofetil
- Warning
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'