Läs mer
Mycophenolic Acid Acyl-Beta-D-glucuronide, TRC
Produktkod. 30137623
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137623 5 mg
30137640 10 mg
18003220 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137623 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM8315255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99043-04-6
Molekylformel C23 H28 O12
Molekylvikt (g/mol) 496.46
Synonym Mycophenolic Acid Acyl-B-D-Glucuronide
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoyl]oxyoxane-2-carboxylic acid
LEDER COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O
Specifikationer
Kemiskt namn eller material Mycophenolic acid acyl-beta-D-glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C23H28O12/c1-9(4-6-11-15(25)14-12(8-33-22(14)31)10(2)19(11)32-3)5-7-13(24)34-23-18(28)16(26)17(27)20(35-23)21(29)30/h4,16-18,20,23,25-28H,5-8H2,1-3H3,(H,29,30)/b9-4+/t16-,17-,18+,20-,23+/m0/s1
Formel vikt 496.1581
Procent renhet >95%