Läs mer
Mycophenolic Acid Phenolic beta-D-Glucoside, TRC
Produktkod. 30137429
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137429 5 mg
30003785 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137429 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM8315305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55533-52-3
Molekylformel C23 H30 O11
Molekylvikt (g/mol) 482.48
Synonym Mycophenolic Acid Phenolic -Ss-D-Glucoside
IUPAC-namn (E)-6-[6-methoxy-7-methyl-3-oxo-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1H-2-benzofuran-5-yl]-5-methylhex-4-enoic acid
LEDER COc1c(C)c2COC(=O)c2c(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)c1C\C(=C\CCC(=O)O)\C
Specifikationer
Kemiskt namn eller material Mycophenolic Acid Phenolic beta-D-Glucoside
Kvantitet 5 mg
InChI formel InChI=1S/C23H30O11/c1-10(5-4-6-15(25)26)7-12-20(31-3)11(2)13-9-32-22(30)16(13)21(12)34-23-19(29)18(28)17(27)14(8-24)33-23/h5,14,17-19,23-24,27-29H,4,6-9H2,1-3H3,(H,25,26)/b10-5+/t14-,17-,18+,19-,23+/m1/s1
Formel vikt 482.1788