missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYD88 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23230-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a cytosolic adapter protein that plays a central role in the innate and adaptive immune response. This protein functions as an essential signal transducer in the interleukin-1 and Toll-like receptor signaling pathways. These pathways regulate that activation of numerous proinflammatory genes. The encoded protein consists of an N-terminal death domain and a C-terminal Toll-interleukin1 receptor domain. Patients with defects in this gene have an increased susceptibility to pyogenic bacterial infections. Alternate splicing results in multiple transcript variants.Specifications
MYD88 | |
Polyclonal | |
Unconjugated | |
MYD88 | |
MYD88, MYD88D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301059, 4615 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6Y1S1, Q99836 | |
MYD88 | |
MYD88 Fusion Protein Ag19770 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |