Learn More
Thermo Scientific™ UV Transilluminator Bulb, 306 nm, for myECL™ Imager
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.
Replacement parts for the myECL Imager and accessories to expand upon its capabilities.
Specifications
Specifications
| Product Type | UV Transilluminator Bulb |
| Description | UV Transilluminator Bulb, 306nm |
| Quantity | 1 bulb |
| For Use With (Equipment) | myECL Imager |
| Product Line | myECL |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
Vous avez repéré une opportunité d'amélioration ?