missing translation for 'onlineSavingsMsg'
Learn More

MYH2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55069-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826515

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to MYH2, will not bind other myosins.

Muscle contraction. Required for cytoskeleton organization.
Specifications

Specifications

MYH2
0.26 mg/mL
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UKX2
MYH2, MYH2A, MYHas8, MyHC 2A, MyHC IIa, MYHSA2, Myosin 2, Myosin 2a, Myosin heavy chain 2, Myosin heavy chain 2a, Myosin heavy chain IIa
Rabbit
IgG
150 μL
-20°C
Primary
4620
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MYH2
Liquid
MYH2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers