missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MYL3 (Human) Recombinant Protein

Human MYL3 full-length ORF ( AAH09790, 1 a.a. - 195 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00004634-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16148081

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MAPKKPEPKKDDAKAAPKAAPAPAPPPEPERPKEVEFDASKIKIEFTPEQIEEFKEAFMLFDRTPKCEMKITYGQCGDVLRALGQNPTQAEVLRVLGKPRQEELNTKMMDFETFLPMLQHISKNKDTGTYEDFVEGLRVFDKEGNGTVMGAELRHVLATLGERLTEDEVEKLMAGQEDSNGCINYEAFVKHIMSS
Specifications

Specifications

AAH09790
myosin, light chain 3, alkali; ventricular, skeletal, slow
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MYL3
GST
4634
Wheat germ expression system
25 μg
CMH8, MLC1SB, MLC1V, VLC1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers