missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ MYL3 Monoclonal Antibody (7C1)

Beskrivning
MA5-15514 targets MYL3 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15514 immunogen is purified recombinant fragment of MYL3 expressed in E. Coli. MA5-15514 detects MYL3 which has a predicted molecular weight of approximately 22kDa.
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifikationer
Specifikationer
| Antigen | MYL3 |
| Användningsområden | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Klassificering | Monoclonal |
| Klona | 7C1 |
| Koncentration | Conc. Not Determined |
| Konjugera | Unconjugated |
| Formulering | ascites with 0.03% sodium azide |
| Gen | MYL3 |
| Genaccessionsnr. | P08590, P16409 |
| Gene Alias | Cardiac myosin light chain 1; CMH8; CMLC1; MLC1s; MLC1SB; Mlc1v; MLClV; MLC-lV/sb; MYL3; Mylc; Mylc1v; Myosin alkali light chain 1, ventricular; Myosin alkali light chain 1, ventricular/slow skeletal muscle isoform; Myosin light chain 1, slow-twitch muscle B/ventricular isoform; myosin light chain 3; Myosin light chain 3, alkali, cardiac ventricles; myosin light chain, alkali, cardiac ventricles; myosin, light chain 3, alkali; ventricular, skeletal, slow; myosin, light polypeptide 3; myosin, light polypeptide 3, alkali; ventricular, skeletal, slow; rVMLC1; slow skeletal; slow skeletal ventricular myosin alkali light chain 3; ventricular; ventricular myosin alkali light chain; Ventricular myosin light chain 1; ventricular, skeletal, slow; ventricular/slow twitch myosin alkali light chain; VLC1; VLCl |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?