missing translation for 'onlineSavingsMsg'
Learn More

MYL5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14249-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16882804

  • 3665.00 SEK / 150µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia.
Specifications

Specifications

MYL5
Polyclonal
Unconjugated
MYL5
MYL5, MyLC 2, MYLC2, Myosin light chain 5
Rabbit
Antigen Affinity Chromatography
RUO
4636
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q02045
MYL5
MYL5 Fusion Protein Ag5522
150 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers