missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYLK2 (skMLCK) Recombinant Human Protein
MYLK2 (skMLCK) is a calcium/calmodulin-dependent serine/threonine kinase.
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_149109 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 92.1kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488513
|
Invitrogen™
PR7110A |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10404213
|
Invitrogen™
PV4025 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MYLK2 (skMLCK) is expressed exclusively in adult skeletal muscle.
Specifications
NP_149109 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MLCK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
92.1kDa | |
RUO - research use only | |
MYLK2 (skMLCK) | |
Human | |
GST Tag | |
Frozen |