missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLK4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24309-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
MYLK4 (Myosin light chain kinase family member 4) is also named as SGK085 and belongs to the CAMK Ser/Thr protein kinase family. MLCKs are activated by Ca2+-calmodulin complexes and require Mg2+ or Ca2+ as a co-factor. It has 2 isoforms produced by alternative splicing.Specifications
MYLK4 | |
Polyclonal | |
Unconjugated | |
MYLK4 | |
MLCK4, MYLK4, SgK085, Sugen kinase 85 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
340156 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86YV6 | |
MYLK4 | |
MYLK4 Fusion Protein Ag19314 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |