missing translation for 'onlineSavingsMsg'
Learn More

MYO9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12432-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840144

  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the myosin family of actin-based molecular motor heavy chain proteins. The protein has four IQ motifs located in the neck domain that bind calmodulin, which serves as a light chain. The protein complex has a single-headed structure and exhibits processive movement on actin filaments toward the minus-end. The protein also has rho-GTPase activity. Polymorphisms in this gene are associated with celiac disease and ulcerative colitis susceptibility. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

MYO9B
Polyclonal
Unconjugated
MYO9B
CELIAC4, MYO9B, MYO9B variant protein, myosin IXB, MYR5
Rabbit
Antigen Affinity Chromatography
RUO
17925, 25486, 4650
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13459, Q63358, Q9QY06
Myo9b
MYO9B Fusion Protein Ag3108
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers