missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoferlin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19548-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The antibody can recognize all the 8 isoforms of MYOF.
Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined.Specifications
Myoferlin | |
Polyclonal | |
Unconjugated | |
MYOF | |
Fer 1 like protein 3, FER1L3, KIAA1207, MYOF, Myoferlin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26509 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZM1 | |
MYOF | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |