missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20360-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to MYOM1.
The giant protein titin, together with its associated proteins, interconnects the major structure of sarcomeres, the M bands and Z discs. The C-terminal end of the titin string extends into the M line, where it binds tightly to M-band constituents of apparent molecular masses of 190 kD (myomesin 1) and 165 kD (myomesin 2). This protein, myomesin 1, like myomesin 2, titin, and other myofibrillar proteins contains structural modules with strong homology to either fibronectin type III (motif I) or immunoglobulin C2 (motif II) domains. Myomesin 1 and myomesin 2 each have a unique N-terminal region followed by 12 modules of motif I or motif II, in the arrangement II-II-I-I-I-I-I-II-II-II-II-II. The two proteins share 50% sequence identity in this repeat-containing region. The head structure formed by these 2 proteins on one end of the titin string extends into the center of the M band. The integrating structure of the sarcomere arises from muscle-specific members of the superfamily of immunoglobulin-like proteins. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
MYOM1 | |
Polyclonal | |
Unconjugated | |
MYOM1 | |
MYOM1, Myomesin 1, myomesin 1, 185kDa, Myomesin family member 1, SKELEMIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17929, 316740, 8736 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52179, Q62234 | |
Myom1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |