missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Light Chain 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5A1F1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Myosin Light Chain 2 |
---|---|
Clone | 5A1F1 |
Concentration | 0.7 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827255
|
Proteintech
60229-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817255
|
Proteintech
60229-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
Myosin Light Chain 2 | |
0.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MLC 2, MLC 2v, MLC2, MLC2v, MYL2, Myosin Light Chain 2 | |
Myl2 | |
IgG1 | |
Protein G | |
Antibody |
5A1F1 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Porcine, Mouse, Rat, Human | |
P08733, P10916, P51667 | |
17906, 363925, 396690, 4633 | |
Myosin Light Chain 2 Fusion Protein Ag1356 | |
Primary | |
-20°C | |
MYL2 |