missing translation for 'onlineSavingsMsg'
Läs mer

MYPN Antibody, Novus Biologicals™

Produktkod. 18156418 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18156418 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18156418 Leverantör Novus Biologicals Leverantörsnummer NBP238924

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

MYPN Polyclonal specifically detects MYPN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYPN
Användningsområden Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassificering Polyclonal
Konjugera Unconjugated
Utspädning Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulering PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Genaccessionsnr. Q86TC9
Gene Alias 145 kDa (MYOP), MYOP145 kDa sarcomeric protein, myopalladin
Gensymboler MYPN
Värd art Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: EDLSNNGSLHSANSTTNLAAIEPQPSPPHSEPPSVEQPPKPKLEGVLVNHNEPRSSSRIGLRVHFNLPEDDKGSEASSE
Reningsmetod Affinity Purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 84665
Testspecificitet Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Målarter Human
Innehåll och lagring Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.