Läs mer
Myristicin Acid Methyl Ester, TRC
Produktkod. 30478651
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478651 100 mg
30470338 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30478651 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM884615100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22934-58-3
Molekylformel C10 H10 O5
Molekylvikt (g/mol) 210.18
Synonym 7-Methoxy-1,3-benzodioxole-5-carboxylic Acid Methyl Ester, 3-Methoxy-4,5-
IUPAC-namn methyl 7-methoxy-1,3-benzodioxole-5-carboxylate
LEDER COC(=O)c1cc(OC)c2OCOc2c1
Specifikationer
Kemiskt namn eller material Myristicin Acid Methyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C10H10O5/c1-12-7-3-6(10(11)13-2)4-8-9(7)15-5-14-8/h3-4H,5H2,1-2H3
Formel vikt 210.0528
Procent renhet >95%