Läs mer
Myristoleic Acid, TRC
Produktkod. 30481493
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30481493 25 mg
1 options
Produktkod. 30481493 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM88460525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Myristoleic acid
CAS 544-64-9
Molekylformel C14 H26 O2
Kvantitet 25 mg
Synonym 9-Tetradecenoic acid','(9Z)- (9CI','ACI), (9Z)-9-Tetradecenoic acid (ACI), 9-Tetradecenoic acid','(Z)- (8CI), (9Z)-Tetradeca-9-enoic acid, (Z)-9-Tetradecenoic acid, 9(Z)-Tetradecenoic acid, 9(Z)-Tetradecenoic acid, 9-cis-Tetradecenoic acid, cis-9-Tetradecenoic acid, cis-Δ9-Tetradecenoic acid, Myristoleic acid, Myristolenic acid, Oleomyristic acid
InChI formel InChI=1S/C14H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h5-6H,2-4,7-13H2,1H3,(H,15,16)/b6-5-
LEDER CCCC\C=C/CCCCCCCC(=O)O
IUPAC-namn (Z)-tetradec-9-enoic acid
Molekylvikt (g/mol) 226.36
Formel vikt 226.1933