Läs mer
(-)-Myrtenol, TRC
Produktkod. 30010136
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010136 10 g
30005736 5 g
30006451 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010136 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM88506310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19894-97-4
Molekylformel C10 H16 O
Molekylvikt (g/mol) 152.23
Synonym (1R,5S)-6,6-Dimethyl-bicyclo(3.1.1)hept-2-ene-2-methanol
IUPAC-namn [(1R,5S)-6,6-dimethyl-4-bicyclo[3.1.1]hept-3-enyl]methanol
LEDER CC1(C)[C@@H]2CC=C(CO)[C@H]1C2
Specifikationer
Kemiskt namn eller material (-)-Myrtenol
Kvantitet 10 g
InChI formel InChI=1S/C10H16O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,8-9,11H,4-6H2,1-2H3/t8-,9-/m1/s1
Formel vikt 152.1201