Läs mer
N-[1,1-Bis[(acetyloxy)methyl]-3-(4-octylphenyl)propyl]acetamide, TRC
Produktkod. 30341707
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30341707 50 mg
1 options
Produktkod. 30341707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB40060050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[1,1-Bis[(acetyloxy)methyl]-3-(4-octylphenyl)propyl]acetamide
CAS 162358-09-0
Molekylformel C25 H39 N O5
Kvantitet 50 mg
Synonym Acetamide, N-[1,1-bis[(acetyloxy)methyl]-3-(4-octylphenyl)propyl]-, 2-Acetamido-2-acetoxymethyl-4-(4-octylphenyl)butyl acetate, Acetic acid 2-acetoxymethyl-2-acetylamino-4-(4-octylphenyl)butyl ester, Triacetylfingolimod, (N,O,O-Triacetyl)fingolimod, N-[1,1-Bis[(acetyloxy)methyl]-3-(4-octylphenyl)propyl]acetamide
InChI formel InChI=1S/C25H39NO5/c1-5-6-7-8-9-10-11-23-12-14-24(15-13-23)16-17-25(26-20(2)27,18-30-21(3)28)19-31-22(4)29/h12-15H,5-11,16-19H2,1-4H3,(H,26,27)
LEDER CCCCCCCCc1ccc(CCC(COC(=O)C)(COC(=O)C)NC(=O)C)cc1
IUPAC-namn [2-acetamido-2-(acetyloxymethyl)-4-(4-octylphenyl)butyl] acetate
Molekylvikt (g/mol) 433.58
Formel vikt 433.2828