Läs mer
N-(1,3,4-Thiadiazolyl)nicotinamide, TRC
Produktkod. 17981555
Change view
Kvantitet:
5 mg
25 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981555 5 mg
17944815 25 mg
17912105 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17981555 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3440005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51987-99-6
Molekylformel C8H6N4OS
Molekylvikt (g/mol) 206.22
Synonym N-(1,3,4-Thiadiazol-2-yl)pyridine-3-carboxamide, N-(1,3,4-Thiadiazolyl)-m-nicotinamide;
LEDER O=C(NC1=NN=CS1)C2=CC=CN=C2
Specifikationer
Kemiskt namn eller material N-(1,3,4-Thiadiazolyl)nicotinamide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C8H6N4OS/c13-7(6-2-1-3-9-4-6)11-8-12-10-5-14-8/h1-5H,(H,11,12,13)
Formel vikt 206.0262
Procent renhet >95%