Läs mer
N-(1-Carboxy-3-phenylpropyl)-S-lisinopril (Mixture of diastereomers), TRC
Produktkod. 30363703
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363703 50 mg
30350779 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC18128050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(1-Carboxy-3-phenylpropyl)-lisinopril
CAS 927819-64-5
Molekylformel C31 H41 N3 O7
Kvantitet 50 mg
Synonym L-Proline, N6-(1-carboxy-3-phenylpropyl)-N2-[(1S)-1-carboxy-3-phenylpropyl]-L-lysyl-, N6-(1-Carboxy-3-phenylpropyl)-N2-[(1S)-1-carboxy-3-phenylpropyl]-L-lysyl-L-proline, N-(1-Carboxy-3-phenylpropyl)-(S)-lisinopril, N-(1-Carboxy-3-phenylpropyl)-lisinopril, (2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-6-[(1-carboxy-3-phenylpropyl)amino]hexanoyl]pyrrolidine-2-carboxylic Acid, N6-(1-Carboxy-3-phenylpropyl)-N2-[(1S)-1-carboxy-3-phenylpropyl]-L-lysyl-L-proline (Mixture of diastereomers)
InChI formel InChI=1S/C31H41N3O7/c35-28(34-21-9-15-27(34)31(40)41)24(33-26(30(38)39)19-17-23-12-5-2-6-13-23)14-7-8-20-32-25(29(36)37)18-16-22-10-3-1-4-11-22/h1-6,10-13,24-27,32-33H,7-9,14-21H2,(H,36,37)(H,38,39)(H,40,41)/t24-,25?,26-,27-/m0/s1
LEDER OC(=O)[C@H](CCc1ccccc1)N[C@@H](CCCCNC(CCc2ccccc2)C(=O)O)C(=O)N3CCC[C@H]3C(=O)O
IUPAC-namn (2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-6-[(1-carboxy-3-phenylpropyl)amino]hexanoyl]pyrrolidine-2-carboxylic acid
Molekylvikt (g/mol) 567.67
Formel vikt 567.2945