Läs mer
N’-(1-Methylazepan-4-yl)benzohydrazine Monohydrochloride Salt, TRC
Produktkod. 30133789
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133789 100 mg
17964295 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133789 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM288685100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 117078-69-0
Molekylformel C14 H21 N3 O . Cl H
Molekylvikt (g/mol) 283.8
Synonym Benzoic acid, 2-(hexahydro-1-methyl-1H-azepin-4-yl)hydrazide, hydrochloride (1:1), Benzoic acid, 2-(hexahydro-1-methyl-1H-azepin-4-yl)hydrazide, monohydrochloride (9CI)
IUPAC-namn N'-(1-methylazepan-4-yl)benzohydrazide;hydrochloride
LEDER Cl.CN1CCCC(CC1)NNC(=O)c2ccccc2
Specifikationer
Kemiskt namn eller material 1-Benzoyl-2-[(4RS)-1-methylhexahydro-1H-azepin-4-yl]diazane Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C14H21N3O.ClH/c1-17-10-5-8-13(9-11-17)15-16-14(18)12-6-3-2-4-7-12;/h2-4,6-7,13,15H,5,8-11H2,1H3,(H,16,18);1H
Formel vikt 283.1451
Procent renhet >95%