Läs mer
N-(1-Naphthyl)phthalamic Acid, TRC
Produktkod. 30467281
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30467281 1 g
1 options
Produktkod. 30467281 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3781151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Naptalam
CAS 132-66-1
Molekylformel C18 H13 N O3
Kvantitet 1 g
Synonym Benzoic acid','2-[(1-naphthalenylamino)carbonyl]- (9CI','ACI), 2-[(1-Naphthalenylamino)carbonyl]benzoic acid (ACI), Phthalamic acid','N-1-naphthyl- (7CI','8CI), 1-Naphthylphthalamic acid, 2-(1-Naphthylcarbamoyl)benzoic acid, 2-(N-(1-Naphthyl)amido)benzoic acid, 2-[(1-Naphthylamino)carbonyl]benzoic acid, Alanap, Alanap 1, Alanap 10G at, Analape, Grelutin, N-1-Naphthylphthalamic acid, N-α-Naphthylphthalamic acid, Naphthalam, Naptalam, NPA, NPA (pesticide), NSC 204421
InChI formel InChI=1S/C18H13NO3/c20-17(14-9-3-4-10-15(14)18(21)22)19-16-11-5-7-12-6-1-2-8-13(12)16/h1-11H,(H,19,20)(H,21,22)
LEDER OC(=O)c1ccccc1C(=O)Nc2cccc3ccccc23
IUPAC-namn 2-(naphthalen-1-ylcarbamoyl)benzoic acid
Molekylvikt (g/mol) 291.3
Formel vikt 291.0895