Läs mer
N-[(1S)-2-Amino-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide, TRC
Produktkod. 30133078
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133078 10 mg
30136800 500 mg
30008121 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133078 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62254010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 289472-80-6
Molekylformel C14 H14 N4 O2
Molekylvikt (g/mol) 270.29
Synonym 2-Pyrazinecarboxamide, N-[(1S)-2-amino-2-oxo-1-(phenylmethyl)ethyl]-, N-[(1S)-2-Amino-1-benzyl-2-oxo-ethyl]pyrazine-2-carboxamide
IUPAC-namn N-[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]pyrazine-2-carboxamide
LEDER NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2
Specifikationer
Kemiskt namn eller material N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide
Analyt- eller komponentnamn N-[(1S)-2-Amino-1-benzyl-2-oxoethyl]pyrazine-2-carboxamide
Frakt skick Room Temperature
InChI formel InChI=1S/C14H14N4O2/c15-13(19)11(8-10-4-2-1-3-5-10)18-14(20)12-9-16-6-7-17-12/h1-7,9,11H,8H2,(H2,15,19)(H,18,20)/t11-/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 10 mg
Formel vikt 270.1117