Läs mer
N-[[2'-(1H-Tetrazol-5-Yl)[1,1'-Biphenyl]-4-Yl]Methyl]Amine, TRC
Produktkod. 30455561
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455561 10 mg
30454449 100 mg
30452873 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455561 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT31092510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147225-68-1
Molekylformel C14H13N5
Molekylvikt (g/mol) 251.29
Synonym 2'-(2H-Tetrazol-5-yl)[1,1'-biphenyl]-4-methanamine, [4-[2-(2H-Tetrazol-5-yl)phenyl]phenyl]methanamine
IUPAC-namn [4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methanamine
LEDER NCc1ccc(cc1)c2ccccc2c3nnn[nH]3
Specifikationer
Kemiskt namn eller material N-((2'-(1H-Tetrazol-5-Yl)(1,1'-Biphenyl)-4-Yl)Methyl)Amine
Kvantitet 10 mg
InChI formel InChI=1S/C14H13N5/c15-9-10-5-7-11(8-6-10)12-3-1-2-4-13(12)14-16-18-19-17-14/h1-8H,9,15H2,(H,16,17,18,19)
Formel vikt 251.1171