Läs mer
N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamide, TRC
Produktkod. 17917904
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17917904 10 mg
17917534 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17917904 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT28396510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 914465-68-2
Molekylformel C19 H21 N5 O
Molekylvikt (g/mol) 335.4
Synonym Pentanamide, N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, Pentanamide, N-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), N-[[2'-(2H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]pentanamide, Valsartan-desvalyl
IUPAC-namn N-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]pentanamide
LEDER CCCCC(=O)NCc1ccc(cc1)c2ccccc2c3nnn[nH]3
Specifikationer
Kemiskt namn eller material N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamide
Kvantitet 10 mg
InChI formel InChI=1S/C19H21N5O/c1-2-3-8-18(25)20-13-14-9-11-15(12-10-14)16-6-4-5-7-17(16)19-21-23-24-22-19/h4-7,9-12H,2-3,8,13H2,1H3,(H,20,25)(H,21,22,23,24)
Formel vikt 335.1746