missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dibromophenyl)-3,5-difluorobenzamide, 97%

Brand:  Alfa Aesar™ H61658.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458218

  • 5659.56 SEK / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(2,4-Dibromophenyl)-3,5-difluorobenzamide
391.01
MFCD15335464

Specifications

N-(2,4-Dibromophenyl)-3,5-difluorobenzamide
97%
MFCD15335464
Solid
391.01
1g
Videos
SDS