missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2,4-Dichlorophenyl)-3,5-difluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62421.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15497416

  • 13699.94 SEK / 5g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

C12H7Cl2F2NO2S
MFCD24849048

Specifications

N-(2,4-Dichlorophenyl)-3,5-difluorobenzenesulfonamide
C12H7Cl2F2NO2S
338.16
5g
MFCD24849048
97%
Videos
SDS