Läs mer
N-(2,4-Dimethylphenyl)formamide-d9, TRC
Produktkod. 30373971
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373971 1 mg
30372194 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30373971 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4767221MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1801691-99-5
Molekylformel C9H2D9NO
Molekylvikt (g/mol) 158.25
Synonym 2,4-Dimethylformanilide-d9, 2,4-Dimethylphenylformamide-d9, BTS 27919-d9, NSC 406488-d9
LEDER [2H]c1c([2H])c(c([2H])c(c1NC=O)C([2H])([2H])[2H])C([2H])([2H])[2H]
Specifikationer
Kemiskt namn eller material N-(2,4-Dimethylphenyl)formamide-d9
Kvantitet 1 mg
InChI formel InChI=1S/C9H11NO/c1-7-3-4-9(10-6-11)8(2)5-7/h3-6H,1-2H3,(H,10,11)/i1D3,2D3,3D,4D,5D
Formel vikt 158.1406