Läs mer
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide, Mikromol™
Produktkod. 30052545
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052545 25 mg
17912866 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052545 Leverantör Mikromol Leverantörsnummer MM1320.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
CAS 5294-61-1
Analyt- eller komponentnamn N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide
Frakt skick Room Temperature
Molekylformel C14 H21 N3 O
Certifikat och efterlevnad ISO 9001
Synonym N-(2,6-Dimethylphenyl)-2-(piperazin-1-yl)acetamide, N-(2,6-Dimethylphenyl)-1-piperazineacetamide, Ranolazine Related Compound C (USP), Ranolazine Related Compound C
InChI formel InChI=1S/C14H21N3O/c1-11-4-3-5-12(2)14(11)16-13(18)10-17-8-6-15-7-9-17/h3-5,15H,6-10H2,1-2H3,(H,16,18)
LEDER Cc1cccc(C)c1NC(=O)CN2CCNCC2
IUPAC-namn N-(2,6-dimethylphenyl)-2-piperazin-1-ylacetamide
Molekylvikt (g/mol) 247.34
Kvantitet 25 mg
Formel vikt 247.1685 g/mol