Läs mer
N-(2-Aminoethyl)-2,2-diphenylcyclopropanecarboxamide, TRC
Produktkod. 30137271
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137271 250 mg
30008791 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137271 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA607890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 109546-07-8
Molekylformel C18 H20 N2 O
Molekylvikt (g/mol) 280.36
Synonym Cibenzoline Impurity (N-(2-aminoethyl-2,2-diphenyl Cyclopropanecarboxamide)
IUPAC-namn N-(2-aminoethyl)-2,2-diphenylcyclopropane-1-carboxamide
LEDER NCCNC(=O)C1CC1(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material N-(2-Aminoethyl)-2,2-diphenylcyclopropanecarboxamide
Kvantitet 250 mg
InChI formel InChI=1S/C18H20N2O/c19-11-12-20-17(21)16-13-18(16,14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-10,16H,11-13,19H2,(H,20,21)
Formel vikt 280.1576