Läs mer
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride), Mikromol™
Produktkod. 17904376
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17904376 25 mg
17961736 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17904376 Leverantör Mikromol Leverantörsnummer MM0075.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride)
CAS 73664-32-1
Analyt- eller komponentnamn N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride (Naphthylacetylethylenediamine Hydrochloride)
Frakt skick Room Temperature
Molekylformel C14 H16 N2 O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Naphazoline Hydrochloride Imp. A (EP), Naphazoline Imp. A (EP), Naphazoline USP Related Compound A, Naphazoline USP RC A, N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide Hydrochloride, Naphazoline Nitrate Imp. A (EP) as Hydrochloride, Naphazoline Hydrochloride Imp. A (EP) as Hydrochloride, Naphthylacetylethylenediamine Hydrochloride, Naphazoline Related Compound A as Hydrochloride, Naphazoline Hydrochloride Impurity A as Hydrochloride, Naphazoline Nitrate Impurity A as Hydrochloride, Naphazoline Impurity A as Hydrochloride, Naphazoline Impurity A as Hydrochloride
InChI formel InChI=1S/C14H16N2O.ClH/c15-8-9-16-14(17)10-12-6-3-5-11-4-1-2-7-13(11)12;/h1-7H,8-10,15H2,(H,16,17);1H
LEDER Cl.NCCNC(=O)Cc1cccc2ccccc12
IUPAC-namn N-(2-aminoethyl)-2-naphthalen-1-ylacetamide;hydrochloride
Molekylvikt (g/mol) 264.75
Kvantitet 25 mg
Formel vikt 264.1029 g/mol