Learn More
Alfa Aesar™ N-(2-Bromo-4-fluorophenyl)-3,5-difluorobenzenesulfonamide, 97%
Brand:  Alfa Aesar™ H62945.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15478026
  • 11319.11 SEK / 5g
Estimated Shipment date
from Supplier 08-03-2018
Add to basket
Specifications
Chemical Identifiers
N-(2-Bromo-4-fluorophenyl)-3,5-difluorobenzenesulfonamide
366.15
MFCD26960685
Specifications
N-(2-Bromo-4-fluorophenyl)-3,5-difluorobenzenesulfonamide
97%
366.15
Solid
5g
MFCD26960685