Learn More
N-(2-Bromobenzyl)acetamide, 97%, Thermo Scientific™
Catalog Number 11450090
Click to view available options
Quantity:
1 g
5 g
74315-07-4
C9H10BrNO
228.09
MFCD00831169
BWFCPQHLMPFBQH-UHFFFAOYSA-N
n-2-bromobenzyl acetamide, n-2-bromophenyl methyl acetamide, maybridge1_005649, acetamide,n-2-bromophenyl methyl
2824953
N-[(2-bromophenyl)methyl]acetamide
CC(=O)NCC1=CC=CC=C1Br
74315-07-4
C9H10BrNO
228.09
MFCD00831169
BWFCPQHLMPFBQH-UHFFFAOYSA-N
n-2-bromobenzyl acetamide, n-2-bromophenyl methyl acetamide, maybridge1_005649, acetamide,n-2-bromophenyl methyl
2824953
N-[(2-bromophenyl)methyl]acetamide
CC(=O)NCC1=CC=CC=C1Br
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 78°C to 80°C
Quantity 1 g
Formula Weight 228.09
Percent Purity 97%
Chemical Name or Material N-(2-Bromobenzyl)acetamide