missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[2-Chloro-5-(trifluoromethyl)phenyl]-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65497.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449686

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-[2-Chloro-5-(trifluoromethyl)phenyl]-2,2-diphenylacetamide
389.8
MFCD00359487

Specifications

N-[2-Chloro-5-(trifluoromethyl)phenyl]-2,2-diphenylacetamide
95%
MFCD00359487
Solid
389.8
1g
Videos
SDS