Läs mer
N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-L-valine Methyl Ester, TRC
Produktkod. 30367240
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30367240 25 mg
1 options
Produktkod. 30367240 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC96237025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-L-valine Methyl Ester
CAS 137863-90-2
Molekylformel C25 H30 N2 O3
Kvantitet 25 mg
Synonym L-Valine, N-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-, methyl ester, N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-N-(1-oxopentyl)-L-valine methyl ester
InChI formel InChI=1S/C25H30N2O3/c1-5-6-11-23(28)27(24(18(2)3)25(29)30-4)17-19-12-14-20(15-13-19)22-10-8-7-9-21(22)16-26/h7-10,12-15,18,24H,5-6,11,17H2,1-4H3/t24-/m0/s1
LEDER CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2C#N)[C@@H](C(C)C)C(=O)OC
IUPAC-namn methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methyl-pentanoylamino]-3-methylbutanoate
Molekylvikt (g/mol) 406.52
Formel vikt 406.2256