Läs mer
N-(2-Cyanoethyl)-L-valine, TRC
Produktkod. 30354725
Change view
Kvantitet:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354725 2.5 g
30360362 1 g
30349504 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30354725 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9810802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51078-49-0
Molekylformel C8H14N2O2
Molekylvikt (g/mol) 170.21
Synonym N-(2-Cyanoethyl)valine
IUPAC-namn (2S)-2-(2-cyanoethylamino)-3-methylbutanoic acid
LEDER CC(C)[C@H](NCCC#N)C(=O)O
Specifikationer
Kemiskt namn eller material N-(2-Cyanoethyl)-L-valine
InChI formel InChI=1S/C8H14N2O2/c1-6(2)7(8(11)12)10-5-3-4-9/h6-7,10H,3,5H2,1-2H3,(H,11,12)/t7-/m0/s1
Kvantitet 2.5 g
Formel vikt 170.1055