Läs mer
N-2-Desmethyl Ropivacaine, TRC
Produktkod. 30352443
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352443 100 mg
30357773 1 g
30357185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352443 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD292380100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98626-59-6
Molekylformel C16H24N2O
Molekylvikt (g/mol) 260.37
Synonym (2S)-N-(2,6-Dimethylphenyl)-1-ethyl-2-piperidinecarboxamide, (S)-N-(2,6-Dimethylphenyl)-1-ethylpiperidine-2-carboxamide, Ropivacaine EP Impurity D
LEDER O=C([C@@H]1N(CC)CCCC1)NC2=C(C)C=CC=C2C
Specifikationer
Kemiskt namn eller material N-2-Desmethyl Ropivacaine
Kvantitet 100 mg
InChI formel InChI=1S/C16H24N2O/c1-4-18-11-6-5-10-14(18)16(19)17-15-12(2)8-7-9-13(15)3/h7-9,14H,4-6,10-11H2,1-3H3,(H,17,19)/t14-/m0/s1
Formel vikt 260.1889