Läs mer
N-(2-Hydroxyethyl)nicotinamide, TRC
Produktkod. 17992185
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17992185 1 g
17911225 250 mg
17950615 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17992185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9423651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6265-73-2
Molekylformel C8 H10 N2 O2
Molekylvikt (g/mol) 166.18
Synonym 3-Pyridinecarboxamide, N-(2-hydroxyethyl)-, Nicotinamide, N-(2-hydroxyethyl)- (6CI,7CI,8CI), N-(2-Hydroxyethyl)-3-pyridinecarboxamide, 3-(2-Hydroxyethyl)carbamoylpyridine, N-(2-Hydroxyethyl)nicotinamide, N-Nicotinoyl-2-aminoethanol, N-Nicotinoylethanolamine, NSC 33142, SG 86, Nicorandil amide
IUPAC-namn N-(2-hydroxyethyl)pyridine-3-carboxamide
LEDER OCCNC(=O)c1cccnc1
Specifikationer
Kemiskt namn eller material Nicorandil Amide
Kvantitet 1 g
InChI formel InChI=1S/C8H10N2O2/c11-5-4-10-8(12)7-2-1-3-9-6-7/h1-3,6,11H,4-5H2,(H,10,12)
Formel vikt 166.0742