missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(2-Isopropylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65575.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15482569

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(2-Isopropylphenyl)-2,2-diphenylacetamide
329.44
MFCD01349298

Specifications

N-(2-Isopropylphenyl)-2,2-diphenylacetamide
Solid
329.44
1g
95%
MFCD01349298
Videos
SDS