Läs mer
[N-(2-Maleimidoethyl]ethylenediamine-N,N,N’,N’-tetraacetic Acid, Monoamide, Technical Grade, TRC
Produktkod. 30475066
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30475066 10 mg
30477663 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30475066 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM13848010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1185102-85-5
Molekylformel C16H22N4O9
Molekylvikt (g/mol) 414.37
Synonym [N-(2-Maleimidoethyl]Ethylenediamine-N,N,N',N'-Tetraacetic Acid, Monoamide
IUPAC-namn 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-(2,5-dioxopyrrolidin-1-yl)ethylamino]-2-oxoethyl]amino]acetic acid
LEDER OC(=O)CN(CCN(CC(=O)O)CC(=O)NCCN1C(=O)CCC1=O)CC(=O)O
Specifikationer
Kemiskt namn eller material [N-(2-Maleimido)ethyl]ethylenediamine-N,N,N',N'-tetraacetic Acid, Monoamide, Technical Grade
Kvantitet 10 mg
InChI formel InChI=1S/C16H24N4O9/c21-11(17-3-4-20-12(22)1-2-13(20)23)7-18(8-14(24)25)5-6-19(9-15(26)27)10-16(28)29/h1-10H2,(H,17,21)(H,24,25)(H,26,27)(H,28,29)
Formel vikt 416.1543