Läs mer
N-[2-(Methacryloyloxy)ethyl]-N,N-dimethyl-N-(3-sulfopropyl)betaine, TRC
Produktkod. 30477159
Change view
Kvantitet:
100 g
Produktkod. Kvantitet
30477159 100 g
1 options
Produktkod. 30477159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM226105100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-[2-(Methacryloyloxy)ethyl]-N,N-dimethyl-N-(3-sulfopropyl)betaine
CAS 3637-26-1
Molekylformel C11 H21 N O5 S
Kvantitet 100 g
Synonym N-(3-Sulfopropyl)-N-(methacryloyloxyethyl)-N,N-dimethylammonium Betaine, N-(3-Sulfopropyl)-N-(methacryloxyethyl)-N,N-dimethylammonium Betaine, N-(2-Methacryloyloxyethyl)-N,N-dimethyl-N-(sulfopropyl)betain, N,N-Dimethyl-N-[2-(methacryloyloxy)ethyl]-N-(3-sulfopropyl)ammonium Betaine, N,N-Dimethyl-N-methacryloyloxyethyl-N-(3-sulfopropyl)ammonium Inner Salt, N,N-Dimethyl-N-[2-(methacryloyloxy)ethyl]-N-(3-sulfopropyl)ammonium Betaine, 3-[Dimethyl(methacryloyloxyethyl)ammonium]propanesulfonate, 3-[N,N-Dimethyl-N-(methacryloyloxyethyl)ammonium]propanesulfonate, DMAPS, Dimethyl(2-methacryloyloxyethyl)(3-sulfopropyl)ammonium, Hartomer HB 6380, N,N-Dimethyl(methacryloyloxyethyl)ammonium propanesulfonate, N,N-Dimethyl-N-(3-sulfonatopropyl)-2-(methacryloyloxy)ethane-1-aminium, N,N-Dimethyl-N-[2-[(2-methyl-1-oxo-2-propenyl)oxy]ethyl]-3-sulfo-1-propanaminium Inner Salt
InChI formel InChI=1S/C11H21NO5S/c1-10(2)11(13)17-8-7-12(3,4)6-5-9-18(14,15)16/h1,5-9H2,2-4H3
LEDER CC(=C)C(=O)OCC[N+](C)(C)CCCS(=O)(=O)[O-]
IUPAC-namn 3-[dimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azaniumyl]propane-1-sulfonate
Molekylvikt (g/mol) 279.353
Formel vikt 279.114