Läs mer
N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine, Mikromol™
Produktkod. 30462569
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462569 4 x 25 mg
30052287 100 mg
30049996 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30462569 Leverantör Mikromol Leverantörsnummer MM0065.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
CAS 80638-08-0
Analyt- eller komponentnamn N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
Frakt skick Room Temperature
Molekylformel C14 H20 N2 O2
Certifikat och efterlevnad ISO 17025
Synonym N-(2-Nitrobenzyl)-N-cyclohexyl-N-methylamine
InChI formel InChI=1S/C14H20N2O2/c1-15(13-8-3-2-4-9-13)11-12-7-5-6-10-14(12)16(17)18/h5-7,10,13H,2-4,8-9,11H2,1H3
Lösningstyp Neat
LEDER CN(Cc1ccccc1[N+](=O)[O-])C2CCCCC2
IUPAC-namn N-methyl-N-[(2-nitrophenyl)methyl]cyclohexanamine
Molekylvikt (g/mol) 248.32
Kvantitet 4 x 25 mg
Formel vikt 248.1525 g/mol