missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Propynyl)-2,3-dihydroinden-1-amine, TRC

Product Code. 30423580
Change view
Click to view available options
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30423580 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423580 Supplier Toronto Research Chemicals Supplier No. TRCP839215500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material rac-Rasagiline
CAS 1875-50-9
Molekylformel C12 H13 N
Kvantitet 500 mg
Synonym N-(2-Propynyl)-2,3-dihydroinden-1-amine, 1H-Inden-1-amine','2,3-dihydro-N-2-propyn-1-yl-, 1-Indanamine','N-2-propynyl- (7CI,8CI), 1H-Inden-1-amine','2,3-dihydro-N-2-propynyl- (9CI), 2,3-Dihydro-N-2-propyn-1-yl-1H-inden-1-amine, AGN 1135, Racemic rasagiline
InChI formel InChI=1S/C12H13N/c1-2-9-13-12-8-7-10-5-3-4-6-11(10)12/h1,3-6,12-13H,7-9H2
LEDER C#CCNC1CCc2ccccc12
IUPAC-namn N-prop-2-ynyl-2,3-dihydro-1H-inden-1-amine
Molekylvikt (g/mol) 171.238
Formel vikt 171.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.