Läs mer
N-(2-Pyridinyl)oxamic Acid, Mikromol™
Produktkod. 17994186
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994186 25 mg
17922336 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17994186 Leverantör Mikromol Leverantörsnummer MM0084.140025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2-Pyridinyl)oxamic Acid
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(2-Pyridinyl)oxamic Acid
CAS 13120-39-3
Analyt- eller komponentnamn N-(2-Pyridinyl)oxamic Acid
Frakt skick Room Temperature
Molekylformel C7 H6 N2 O3
Certifikat och efterlevnad ISO 9001
Synonym N-(2-Pyridinyl)oxamic Acid
InChI formel InChI=1S/C7H6N2O3/c10-6(7(11)12)9-5-3-1-2-4-8-5/h1-4H,(H,11,12)(H,8,9,10)
LEDER OC(=O)C(=O)Nc1ccccn1
IUPAC-namn 2-oxo-2-(pyridin-2-ylamino)acetic acid
Molekylvikt (g/mol) 166.13
Kvantitet 25 mg
Formel vikt 166.0378 g/mol