Läs mer
N-(2-Pyridinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide (Flecainide Impurity), TRC
Produktkod. 30425906
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30425906 2.5 mg 2.5mg
30426655 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425906 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP9916902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57415-36-8
Molekylformel C17H14F6N2O3
Molekylvikt (g/mol) 408.3
Synonym Benzamide, N-(2-pyridinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)-, N-(2-Pyridinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide, 2,5-Bis(2,2,2-trifluoroethoxy)-N-(2-pyridylmethyl)benzamide, N-(Pyridin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
LEDER FC(F)(F)COc1ccc(OCC(F)(F)F)c(c1)C(=O)NCc2ccccn2
Specifikationer
Kemiskt namn eller material N-(2-Pyridinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
Kvantitet 2.5 mg
InChI formel InChI=1S/C17H14F6N2O3/c18-16(19,20)9-27-12-4-5-14(28-10-17(21,22)23)13(7-12)15(26)25-8-11-3-1-2-6-24-11/h1-7H,8-10H2,(H,25,26)
Formel vikt 408.0909