Läs mer
N-(2-Succinyl) Phenylephrine(Mixture of Diastereomers), TRC
Produktkod. 30410149
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30410149 5 mg
1 options
Produktkod. 30410149 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6888505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material N-(2-Succinyl) Phenylephrine (Mixture of Diastereomers)
CAS 915278-80-7
Molekylformel C13 H17 N O6
Kvantitet 5 mg
Synonym N-[(2R)-2-Hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methyl-aspartic Acid
InChI formel InChI=1S/C13H17NO6/c1-14(10(13(19)20)6-12(17)18)7-11(16)8-3-2-4-9(15)5-8/h2-5,10-11,15-16H,6-7H2,1H3,(H,17,18)(H,19,20)/t10?,11-/m0/s1
LEDER CN(C[C@H](O)c1cccc(O)c1)C(CC(=O)O)C(=O)O
IUPAC-namn 2-[[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-methylamino]butanedioic acid
Molekylvikt (g/mol) 283.28
Formel vikt 283.1056