missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[2-(Trifluoromethyl)phenyl]-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65747.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479866

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

N-[2-(Trifluoromethyl)phenyl]-5,6,7,8-tetrahydronaphthalene-1-carboxamide
319.33
MFCD27975812

Specifications

N-[2-(Trifluoromethyl)phenyl]-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
319.33
1g
95%
MFCD27975812
Videos
SDS