Läs mer
N-[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]-L-aspartic Acid, TRC
Produktkod. 30179478
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179478 100 mg
30179123 10 mg
30179319 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD790020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 860295-20-1
Molekylformel C13H13NO7
Molekylvikt (g/mol) 295.24
Synonym N-[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]-L-aspartic Acid
LEDER OC(=O)C[C@H](NC(=O)\C=C\c1ccc(O)c(O)c1)C(=O)O
Specifikationer
Kemiskt namn eller material N-[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]-L-aspartic Acid
InChI formel InChI=1S/C13H13NO7/c15-9-3-1-7(5-10(9)16)2-4-11(17)14-8(13(20)21)6-12(18)19/h1-5,8,15-16H,6H2,(H,14,17)(H,18,19)(H,20,21)/b4-2+/t8-/m0/s1